CAP SNAP 11MM CLEAR POLYETHYLENE INTEGRAL PE SEPTUM THINNED PENETRATION AREA 100/PACK 1 * 100 items
Catalog # HYPE6ERC11PEC
Supplier: Thermo Fisher Scientific
CAP SNAP 11MM CLEAR POLYETHYLENE INTEGRAL PE SEPTUM THINNED PENETRATION AREA 100/PACK 1 * 100 items
Catalog # HYPE6ERC11PEC
Supplier: Thermo Fisher Scientific
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR® Blender Bags
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Caps for 2 ml polypropylene snap top microvials for <2 ml samples, level 1 everyday analysis, SureSTART™
Supplier: Thermo Fisher Scientific
Polypropylene snap top 2 ml microvials for <2 ml samples, level 1 everyday analysis, SureSTART™
Supplier: Thermo Fisher Scientific