ContactUsLinkComponent
5 components: 100mg/l each of As Ni Pb Sb Si in HNO3 3% HCl 9% 1 * 500 mL
N° de catalogue CMPLRM096447L5
5 components: 100mg/l each of As Ni Pb Sb Si in HNO3 3% HCl 9% 1 * 500 mL
N° de catalogue CMPLRM096447L5
Frequently Bought Together