Order Entry
Belgium
Orders LinkContactUsLinkComponent
Control Expansion

Control Expansion

RNA control expansion, 6, 1 KIT
N° de catalogue OXNTEXP-RCS001
Control Expansion
N° de catalogue OXNTEXP-RCS001
Numéro CAS:  
Control Expansion
Control Expansion

RNA control expansion, 6, 1 KIT

Numéro CAS:  
Prix
64,80 €
Connexion Pour la disponibilité
1 * 1 KIT
Product Details & Documents

DNA or RNA for sequencing control experiments.

  • Perform DNA or RNA control experiments to familiarise yourself with nanopore sequencing

DNA control expansion (EXP-CTL001): Lambda DNA is 48 kb long and serves as a good model system to evaluate the nanopore sequencing workflow. Control experiments should be carried out when users are getting started with nanopore sequencing devices, or when troubleshooting library preparation. Pack of six vials. Contains Lambda DNA (LMD), sufficient for at least 24 reactions.

RNA control expansion (EXP-RCS001): contains RNA control sample allowing control runs to support Oxford Nanopore cDNA sequencing kits. Pack of three vials. Contains sufficient control RNA for six reactions.

Livraison: Kits are shipped at 2 to 8 °C in an environmentally-friendly temperature-controlled shipping box. Upon receipt, please place the product in a long-term storage location at −20 °C. Oxford Nanopore Technologies deem the useful life of the product to be 3 months from receipt by the customer.

Attention: Oxford Nanopore Technologies products are not intended for use for health assessment or to diagnose, treat, mitigate, cure, or prevent any disease or condition.

Spécifications

Spécifications

  • Pk:
    1 KIT
  • Description:
    RNA control expansion
  • No. of reactions:
    6

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.