Neutralized Sodium hypochlorite 0.25% solution, attached activator, SimpleMix, 16 oz Attached Trigger Spray 1 * 12 Can
Catalog # VELTSHC-NPH.25-16Z
Supplier: VELTEK ASSOCIATES
Neutralized Sodium hypochlorite 0.25% solution, attached activator, SimpleMix, 16 oz Attached Trigger Spray 1 * 12 Can
Catalog # VELTSHC-NPH.25-16Z
Supplier: VELTEK ASSOCIATES
Frequently Bought Together
Methyltrichlorosilane for synthesis, Sigma-Aldrich®
Supplier: Merck
Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals
N,N-Dimethylformamide ≥99.9%, CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dimethyl sulphoxide ≥99.9% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Water, Ultrapure for HPLC
Supplier: Thermo Fisher Scientific
Ethyl acetate 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Heptafluorobutyric anhydride (HFAA), Supelco®
Supplier: Merck
Methanol, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
1-Propanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethyl acetate ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
Benzyl alcohol 99%, pure
Supplier: Thermo Fisher Scientific
Water, pure, demineralised
Supplier: Thermo Fisher Scientific
Diisopropyl ether stabilised for analysis
Supplier: Thermo Fisher Scientific
Hexane (mixture of isomers) ≥98.5%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals