ContactUsLinkComponent
BIOne 1250 Controller with Right Sided Configuration. Includes 4 Pumps and Touch Screen Display. Does Not Included Gas Control, Vessel, or Probes. 230V 1 * 1 items
Catalog # DISI2022-8121
Supplier:  DISTEK
BIOne 1250 Controller with Right Sided Configuration. Includes 4 Pumps and Touch Screen Display. Does Not Included Gas Control, Vessel, or Probes. 230V 1 * 1 items
Catalog # DISI2022-8121
Supplier:  DISTEK
Frequently Bought Together