Orders LinkContactUsLinkComponent
VWR®, Cell Holders
VWR®, Cell Holders
Auto cell holder, 5-position 10 - 100 mm cell holder (for VWR® P4, PV4, M4, P6, M6)
Catalog # 634-1233
Supplier:   VWR Collection
VWR®, Cell Holders
Catalog # 634-1233
Supplier:  VWR Collection
VWR®, Cell Holders
VWR®, Cell Holders
Auto cell holder, 5-position 10 - 100 mm cell holder (for VWR® P4, PV4, M4, P6, M6)
Price
1 060,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Auto cell holder, 5-position 10 - 100 mm cell holder (for VWR® P4, PV4, M4, P6, M6)
  • Seq:
    10