APESIN DAILY BELGIUM (1603 B) 5L 1 * 2 items
Catalog # TANA713195
Fournisseur: WERNER MERTZ
Frequently Bought Together
Xylene (mixture of isomers) ≥98%, TECHNICAL
Fournisseur: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Fournisseur: VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Fournisseur: VWR Chemicals
Tetrahydrofuran, Uvasol® for spectroscopy, Supelco®
Fournisseur: Merck
Methanol ≥99.9%, EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®
Fournisseur: Merck
Acetone ≥99.5%, GPR RECTAPUR®
Fournisseur: VWR Chemicals
Cyclohexane, LiChrosolv® for HPLC, Supelco®
Fournisseur: Merck
Acetone ≥99%, TECHNICAL
Fournisseur: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Fournisseur: VWR Chemicals
2,2,4-Trimethylpentane, Uvasol® for spectroscopy, Supelco®
Fournisseur: Merck
Ethyl acetate ≥99%, GPR RECTAPUR®
Fournisseur: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Fournisseur: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
Fournisseur: VWR Chemicals
Ethanol, HiPerSolv CHROMANORM® for HPLC
Fournisseur: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Fournisseur: VWR Chemicals
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
Fournisseur: VWR Chemicals