0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
50 TABLETS NICKEL HR N° 2 PHOTOMETER 1 * 50 items
WTIDTBSPNICKHR250
:
WATER ID PRIMELAB
50 TABLETS NICKEL HR N° 2 PHOTOMETER 1 * 50 items
WTIDTBSPNICKHR250
:
WATER ID PRIMELAB
Frequently Bought Together
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
: Merck
Test kits, phosphate, Spectroquant®, Supelco®
: Merck
Immersion oil for microscopy, Sigma-Aldrich®
: Merck
Sodium hydroxide, pellets, pure, Supelco®
: Merck
pH indicator strips, MQuant®, Supelco®
: Merck
Test kits, ammonium, Spectroquant®, Supelco®
: Merck
Test kits, nitrate, Spectroquant®, Supelco®
: Merck
Test strips for peroxide, MQuant®, Supelco®
: Merck
pH indicator papers, MQuant®, Supelco®
: Merck
Test kits, nitrogen (total) cell tests, Spectroquant®, Supelco®
: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
: Merck
VWR® Dosatest, pH indicator test strips, non bleeding
: VWR Chemicals