2ML PL UREA 2 (50 TESTS) UREA PHOTOMETER 1 * 2 mL
Catalog # WTIDPOL2UREA2
Supplier: WATER ID PRIMELAB
2ML PL UREA 2 (50 TESTS) UREA PHOTOMETER 1 * 2 mL
Catalog # WTIDPOL2UREA2
Supplier: WATER ID PRIMELAB
Frequently Bought Together
Test strips for peracetic acid, MQuant®, Supelco®
Supplier: Merck
Test strips, Glucose, QUANTOFIX®
Supplier: MACHEREY-NAGEL
Titrimetric rapid tests, hardness, VISOCOLOR®
Supplier: MACHEREY-NAGEL
Sodium hydroxide 1 mol/l, EMPROVE® EXPERT, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Chinese ink, blue, Q Path® stain for histology
Supplier: VWR Chemicals
Colorimetric rapid tests, phosphate, VISOCOLOR®
Supplier: MACHEREY-NAGEL
Ammonia solution 20%, GPR RECTAPUR®
Supplier: VWR Chemicals
Trifluoroacetic acid ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
MHC DEXTRAMER HLAA0201/AIQDLCLAV 1 * 50 Tests
Supplier: MARKET SOURCE PART PROCESS
[EN]CAUSTIC CLEANING SOLUTION 1 * 500 mL
Supplier: PROLABO CMP
Tetrabutylammonium hydrogen sulphate 99% for HPLC
Supplier: Thermo Fisher Scientific
Tartrazine ≥85% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Reagent Repl.Alkalinity M (Alkaphot M) 1 * 250 items
Supplier: Palintest
[EN]STANDARD MORPHOLINE 1000PPM IN H2O 1 * 500 mL
Supplier: PROLABO CMP
Chemkey® Reagents
Supplier: Hach