REAGENT KIT FOR PRIMELAB ID87 (81 TESTS) 1 * 1 items
Catalog # WTIDPL87-KIT
Supplier: WATER ID PRIMELAB
REAGENT KIT FOR PRIMELAB ID87 (81 TESTS) 1 * 1 items
Catalog # WTIDPL87-KIT
Supplier: WATER ID PRIMELAB
Frequently Bought Together
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Photometric test kits, COD VARIO tube tests
Supplier: Lovibond
L(+)-Cysteine hydrochloride monohydrate 98.5-101.0% (alkalimetric, calculated on anhydrous substance), EMPROVE® ESSENTIAL Ph. Eur., USP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Ammonia 25% (as NH3) (BP, Ph. Eur.) pure, pharma grade 1 * 1.000 mL
Supplier: PanReac AppliChem
Test kits, photometric, ammonium, LCK cuvette tests
Supplier: Hach
Powder pillows, reagents for Hach instruments
Supplier: Hach
Histopaque®, sterile filtered, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Methylene blue hydrate, high purity biological stain
Supplier: Thermo Fisher Scientific
HEMATOXYLIN SOLUTION GILL III SURGIPATH 1 * 1 L
Supplier: LEICA HISTOLOGY
EOSIN RAL 555 2.5L 1 * 2,5 L
Supplier: RAL
Reagents for ammonia determination using Lovibond® MD photometers
Supplier: Lovibond
Chemkey® Reagents
Supplier: Hach
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach
NEISSER´S SOLUTION III FOR MICROSCOPY 1 * 100 mL
Supplier: Roth Carl