65 ML PL CHLORIDE 2 (108 TESTS) 1 * 65 mL
Catalog # WTIDPL65CHLORIDE2
Supplier: WATER ID PRIMELAB
Frequently Bought Together
Chromeazurol S (Mordant blue 29), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Reagents for Phosphate Determination using Lovibond® MD photometers
Supplier: Lovibond
BIOTECTOR 1.8N SULFURIC ACID 80MG/L MN 1 * 20 L
Supplier: Hach
Test kits, drop count tests/
Supplier: Hach
Powder pillows, reagents for Hach instruments
Supplier: Hach
[EN]IGG4 ISOTYPE CONTROL S228P/R409K 1 * 1 mg
Supplier: MARKET SOURCE PART PROCESS
SODIUM HYDROXIDE 0.005N 1 * 1 L
Supplier: PROLABO CMP
MOUSE IFN-Α CALIBRATOR 1 * 1 Vial
Supplier: MESO SCALE DISCOVERY
Sulphuric acid ≥96% VLSI for microelectronic, J.T.Baker®
Supplier: Avantor
TITRATION SOLUTION DUROVAL C 1 * 50 ml
Supplier: Heyl GmbH & Co, Gebr.
Diluid III Diff for hematology, J.T.Baker®
Supplier: Avantor