REAGENT KIT FOR PRIMELAB ID103(108TESTS) 1 * 1 items
Catalog # WTIDPL103-KIT
Supplier: WATER ID PRIMELAB
REAGENT KIT FOR PRIMELAB ID103(108TESTS) 1 * 1 items
Catalog # WTIDPL103-KIT
Supplier: WATER ID PRIMELAB
Frequently Bought Together
Test strips, ascorbic acid, Reflectoquant®, Supelco®
Supplier: Merck
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Test strips for ascorbic acid, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide ≥98%, pellets, GPR RECTAPUR®
Supplier: VWR Chemicals
Test strips, peracetic acid, Dosatest
Supplier: VWR Chemicals
Ammonium acetate ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Ammonia solution 28%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Hydrogen peroxide 6% (w/v) (20 volumes), GPR RECTAPUR®
Supplier: VWR Chemicals
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
Supplier: VWR Chemicals
Sodium hypochlorite (12% Cl₂) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Aqueous solution containing sulphuric acid (0.001M) and Triton X-100 (0.225% w/v) 1 * 10 L
Supplier: CUSTOM MADE CHEMICALS
Test kits, photometric, phosphate, LCK cuvette tests
Supplier: Hach
Turbidity standards for 2100 AN and 2100 AN IS
Supplier: Hach
Test kits, photometric, Sulphate, LCK cuvette tests
Supplier: Hach