REAGENT KIT FOR PRIMELAB ID08(50 TESTS) 1 * 1 items
WTIDPL08-KIT
Frequently Bought Together
Colorimetric rapid tests, nitrite, VISOCOLOR®
: MACHEREY-NAGEL
Aqueous solution containing sulphuric acid (0.001M) and Triton X-100 (0.225% w/v) 1 * 10 L
: CUSTOM MADE CHEMICALS
[EN]H2SO4+CITRIC ACID+ISOPROPANOL 1 * 2,5 L
: PROLABO CMP
Histopaque®, sterile filtered, Sigma-Aldrich®
: SIGMA ALDRICH MICROSCOPY
METHANOL LC/MS WITH 0.1% FORMIC ACID 1 * 2,5 L
: PROLABO CMP
Sodium hypochlorite 4-6% available chlorine in aqueous solution
: Thermo Fisher Scientific
pH indicators with integral comparison strips, Whatman™
: Whatman products (Cytiva)
Test strips, dialysis, peroxides, DiaQuant®
: MACHEREY-NAGEL
Paraffin, Solidification point: 56 - 58 °C, pellets for clinical diagnosis
: PanReac AppliChem