ContactUsLinkComponent
The Mag-Bind® Blood & Tissue DNA HDQ 96 Kit is designed for isolation of high-quality genomic DNA from blood samples, saliva, swabs, mouse tails, dried blood spots or cultured cells. 24*96(2304)tests 1 * 1 KIT
Catalog # OMEGM6399-02
Supplier:  OMEGA BIO-TEK
The Mag-Bind® Blood & Tissue DNA HDQ 96 Kit is designed for isolation of high-quality genomic DNA from blood samples, saliva, swabs, mouse tails, dried blood spots or cultured cells. 24*96(2304)tests 1 * 1 KIT
Catalog # OMEGM6399-02
Supplier:  OMEGA BIO-TEK
Frequently Bought Together