NDF SOLUTION 1 * 20 L
Catalog # PRLSMC305320.9020
Supplier: PROLABO CMP
Frequently Bought Together
Soda lime, AnalaR NORMAPUR® analytical reagent, with indicator
Supplier: VWR Chemicals
Test kits, nitrite, Spectroquant®, Supelco®
Supplier: Merck
Test kits, surfactants, Spectroquant®, Supelco®
Supplier: Merck
Test strips, sulphite, MQuant®, Supelco®
Supplier: Merck
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Visual colour kits, ammonium, MQuant®, Supelco®
Supplier: Merck
Ammonia solution 5%
Supplier: PROLABO CMP
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
Supplier: VWR Chemicals
Test kits, manganese, Spectroquant®, Supelco®
Supplier: Merck
Test strips, peroxide, QUANTOFIX®
Supplier: MACHEREY-NAGEL
Sulphate test strips, Dosatest, VWR®
Supplier: VWR Chemicals
Orthoboric acid 40 g/l in aqueous solution for Kjeldahl determinations, with indicator
Supplier: VWR Chemicals
Powder pillows, chlorine, free and total
Supplier: Hach
Test kits, photometric, copper, LCK cuvette tests
Supplier: Hach
VWR® Q Path® Easyfix, Cytology Fixative
Supplier: VWR Chemicals
Reagents for copper determination using Lovibond® MD photometers
Supplier: Lovibond
4 COMPONENTS MIX 1 * 100 mL
Supplier: CUSTOM MADE CHEMICALS LAB
Aqueous solution containing sulphuric acid (0.001M) and Triton X-100 (0.225% w/v) 1 * 10 L
Supplier: CUSTOM MADE CHEMICALS
V-PLEX PROINFLAMMATORY PANEL 1 (HUMAN) KIT 5 PLATES 1 * 1 KIT
Supplier: MESO SCALE DISCOVERY