Lactose (anhydrous) CAS:63-42-3 EC:200-559-2 17034 1 * 5 g
Catalog # CMPLSB48074.5G
Supplier: CUSTOM MADE CHEMICALS LAB
Lactose (anhydrous) CAS:63-42-3 EC:200-559-2 17034 1 * 5 g
Catalog # CMPLSB48074.5G
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Test kits, ammonium, Spectroquant®, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Reagents for nitrate determination using Lovibond® MD photometers
Supplier: Lovibond
NANOCOLOR® Photometric Standard Test Kit for Silica
Supplier: MACHEREY-NAGEL
BACTIDROP CALCOFLUOR WHITE 0.75 ML/AMP. 1 * 50 Ampoul
Supplier: THERMO FISHER DIAGNOSTICS
Bromocresol green-methyl red solution mixed indicator, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Oil Red EGN, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Acid red 52, Technical Grade, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Sodium hydroxide 30% in water, Technical Grade
Supplier: Thermo Fisher Scientific
Nitrite VHR L, 500 ml
Supplier: Lovibond
Direct Red 23, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Aqueous solution containing sulphuric acid (0.001M) and Triton X-100 (0.225% w/v) 1 * 10 L
Supplier: CUSTOM MADE CHEMICALS
Acid Red 183 30% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Fast Red RC Salt, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Potassium hydroxide 85.0-100.5%, pellets Ph. Eur.
Supplier: VWR Chemicals
Colorimetric rapid tests, copper, VISOCOLOR®
Supplier: MACHEREY-NAGEL