ContactUsLinkComponent
7 components: 100mg/l each of Fe Al Cu Zn Ba Mn Cr in HNO3 2% CRM, ISO 17034 and ISO 17025,,,, Conservation 24 months. (12 months after opening),,,, Store under normal laboratory conditions, at temperatures between 15C to 25C,,,, 1 * 100 mL
Catalog # CMPLRM093370L1
7 components: 100mg/l each of Fe Al Cu Zn Ba Mn Cr in HNO3 2% CRM, ISO 17034 and ISO 17025,,,, Conservation 24 months. (12 months after opening),,,, Store under normal laboratory conditions, at temperatures between 15C to 25C,,,, 1 * 100 mL
Catalog # CMPLRM093370L1
Frequently Bought Together