ContactUsLinkComponent
Fractionating columns, Vigreux, w. glass jacket, cone ST 14/23, socket ST 14/23, length: 200 mm, indentations: 15 1 * 1 items
Catalog # WITG1.520.200
Supplier:  witeg Labortechnik
Fractionating columns, Vigreux, w. glass jacket, cone ST 14/23, socket ST 14/23, length: 200 mm, indentations: 15 1 * 1 items
Catalog # WITG1.520.200
Supplier:  witeg Labortechnik
Frequently Bought Together