CIS-RESVERATROL 1 * 10 mg
Catalog # PROOTRCR150005-10
Supplier: LGC Standards PROMOCHEM
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
VWR®, Phosphate 1,000 mg/L in Water standard for ion chromatography
Supplier: VWR Chemicals
pH buffer solutions
Supplier: Mettler - Toledo
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Sulphate 1000 mg/L in water standard for ion chromatography
Supplier: VWR Chemicals
Conductivity standards, KCl, referenced at 25 °C
Supplier: Thermo Fisher Scientific
pH buffer solutions, colour coded
Supplier: Hach
pH buffer, single-use, SINGLET™
Supplier: Hach