Orders LinkContactUsLinkComponent
Bioanalyzer DNA Analysis
Bioanalyzer DNA Analysis
Agilent DNA 1000 reagents, Reagents only, no chips included, sufficient for 25 chips
Catalog # AGLS5067-1505
Supplier:   AGILENT
Bioanalyzer DNA Analysis
Catalog # AGLS5067-1505
Supplier:  AGILENT
Bioanalyzer DNA Analysis
Agilent DNA 1000 reagents, Reagents only, no chips included, sufficient for 25 chips
Price
523,06 €
Log in for availability
1 * 1 ST
Product Details & Documents
DNA fragment or NGS library analysis for sample quality control - for samples up to 12000 bp.
  • High sensitivity from laser (LIF) detection of DNA fragments down to 0,1 ng
  • The broad linear dynamic range allows detection of weak bands without saturation from abundant fragments
  • Sizing and quantitation accuracy and reproducibility results from pre-packaged reagents, standardised assays and automated data analysis
  • Minimal sample consumption is required as only 1 µl is needed for the assay, saving precious material for downstream experiments
  • Fast results are provided, with the analysis of 12 samples performed in only 30 minutes (approximately)
  • Safety is ensured with minimal exposure to hazardous materials, such as ethidium bromide, as only minimal reagents are used
Perform highly accurate and precise DNA electrophoresis with the Bioanalzser DNA analysis solutions. Together with the 2100 Bioanalyzer instrument, the DNA kits enable the analysis of next-generation sequencing libraries and high resolution multiplex PCR reactions, measuring precise size and concentration of all DNA fragments and smears. A large linear dynamic range allows detection of small impurities in PCR purifications.
Besides sample quality control of next-generation sequencing (NGS) libraries, additional research applications for the Bioanalyzer DNA assays include PCR amplicon analysis, multiplex PCR amplicons, gene expression analysis by RT-PCR, restriction fragment analysis, and the detection of targeted cleavage in gene editing studies.
Caution: For research use only. Not for use in diagnostic procedures.
Specifications
  • Description:
    Agilent DNA 1000 reagents
  • Seq:
    04
  • Details:
    Reagents only, no chips included, sufficient for 25 chips