ContactUsLinkComponent
With this frame you can install the Dispensers (PALBGD-4060) to be installed above the cabinets (PALBGDC-4040) without having to mount them to the wall. Creates a free-standing dispenser and cabinet unit. Material of manufacture: 304 stainless steel, 1 * 1 items
Catalog # PALBGDC4040-FRAME
Supplier:  Palbam
With this frame you can install the Dispensers (PALBGD-4060) to be installed above the cabinets (PALBGDC-4040) without having to mount them to the wall. Creates a free-standing dispenser and cabinet unit. Material of manufacture: 304 stainless steel, 1 * 1 items
Catalog # PALBGDC4040-FRAME
Supplier:  Palbam
Frequently Bought Together