XBridge Peptide BEH C18 XP Column, 300Å, 2.5 µm, 4.6mm X 150 mm 1 * 1 items
Catalog # WATE186009076
Supplier: WATERS
XBridge Peptide BEH C18 XP Column, 300Å, 2.5 µm, 4.6mm X 150 mm 1 * 1 items
Catalog # WATE186009076
Supplier: WATERS
Frequently Bought Together
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
Avantor® ACE® C18, HPLC Columns, 5 µm
Supplier: Avantor
Precision screw caps, 18 mm, level 3 high performance applications, SureSTART™
Supplier: Thermo Fisher Scientific
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
VWR®, Crimp Caps for Crimp Neck Headspace Vials ND20
Supplier: VWR Collection
Gold standard syringes for autosampler
Supplier: Agilent
Exhaust filters, V3.0
Supplier: S.C.A.T.
Silica gel 60, irregular 40 - 63 µm for flash chromatography
Supplier: VWR Chemicals
VWR®, Snap Cap Vials
Supplier: VWR Collection
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
Crimp neck vials, N 20
Supplier: MACHEREY-NAGEL
Florisil® 60-100 mesh for chromatography
Supplier: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection