ContactUsLinkComponent
BioResolve RP mAb Polyphenyl Column, 450Å, 2.7 µm, 4.6 mm X 100 mm, with Intact mAb and mAb Subunit Standards 1 * 1 items
Catalog # WATE176004168
Supplier:  WATERS
BioResolve RP mAb Polyphenyl Column, 450Å, 2.7 µm, 4.6 mm X 100 mm, with Intact mAb and mAb Subunit Standards 1 * 1 items
Catalog # WATE176004168
Supplier:  WATERS
Frequently Bought Together