ACQUITY UPLC Peptide HSS T3 C18 Column, 100Å, 1.8 µm, 2.1 mm x 150 mm, with Cytochrome C Digestion Standard 1 * 1 items
Catalog # WATE176003997
Supplier: WATERS
ACQUITY UPLC Peptide HSS T3 C18 Column, 100Å, 1.8 µm, 2.1 mm x 150 mm, with Cytochrome C Digestion Standard 1 * 1 items
Catalog # WATE176003997
Supplier: WATERS
Frequently Bought Together
Accessories for connection system GLS 80
Supplier: DWK Life Sciences
VWR® ACE® Excel® C18 HPLC/UHPLC Columns, Analytical, 2 µm
Supplier: Avantor
Screw neck vials, N 18 and N 20
Supplier: MACHEREY-NAGEL
Solid phase extraction cartridges, Bond Elut™ SI
Supplier: VARIAN
CUSTOM QUANTITAIVE HPLC ANALYSIS 1 * 1 items
Supplier: Avantor
BENSON BP-800 CA 9µM 300 X 7.8MM 1 * 1 items
Supplier: HICHROM
QuEChERS mixes, clean-up, CHROMABOND®
Supplier: MACHEREY-NAGEL
HPLC column tubing
Supplier: Restek
VWR®, Screw Caps for Screw Neck Vials ND10
Supplier: VWR Collection
ELECTRODE ASSEMB. STANDARD FOR G7100ONLY 1 * 1 items
Supplier: Agilent
Size exclusion chromatography columns, Bio SEC-3 and Bio SEC-5
Supplier: Agilent
Safety Waste Caps, Electrically Conductive
Supplier: S.C.A.T.
Avantor® ACE® Excel® C18-Amide, HPLC/UHPLC Columns, 2 µm
Supplier: Avantor
ASTEC CHIROBIOTIC T 5UM 15CM X 4.6MM 1 * 1 Each
Supplier: Merck
GC septa, Thermogreen® LB-2
Supplier: Merck