PLASMA - CELIAC DISEASE (CED), 2ML, CRYOPRESERVED 1 * 2 mL
Catalog # PB006C-1-CED
PLASMA - CELIAC DISEASE (CED), 2ML, CRYOPRESERVED 1 * 2 mL
Catalog # PB006C-1-CED
Frequently Bought Together
Culture media bottles, square, sterile, Nalgene®
Supplier: Thermo Fisher Scientific
VWR®, Cell Strainers
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
Supplier: EPPENDORF
Cryoboxes, Nalgene®
Supplier: Thermo Fisher Scientific
Test tubes
Supplier: Greiner Bio-One
VWR®, Spreaders, T-shaped
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Serological pipettes, Stripette™
Supplier: Corning
Individually Wrapped Serological Pipettes, Falcon®
Supplier: Corning
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
Centrifuge tubes, sterile, polypropylene, Corning®
Supplier: Corning
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Freestanding, with Flat Screw Caps
Supplier: VWR Collection
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF