SARS-COV2 S (424-430) PEPTIDE 1MG, CRYOPRESERVED 1 * 1 mg
Catalog # ANTIGEN-1148
SARS-COV2 S (424-430) PEPTIDE 1MG, CRYOPRESERVED 1 * 1 mg
Catalog # ANTIGEN-1148
Frequently Bought Together
VWR® Multiwell Cell Culture Plates
Supplier: VWR Collection
Media bottles, square, sterile, Nalgene®
Supplier: Thermo Fisher Scientific
VWR®, Stoppers, PE
Supplier: VWR Collection
Serological pipettes, Stripette™
Supplier: Corning
VWR®, Cell Culture Flasks, Non Treated or Treated
Supplier: VWR Collection
Cell culture flasks with standard or filter caps, CELLSTAR®
Supplier: Greiner Bio-One
Pasteur pipettes
Supplier: LP ITALIANA
Microcentrifuge tubes, 0,6/1,5/2,0 ml, graduated, with snap cap
Supplier: Thermo Fisher Scientific
Colour coders for cryovials, CryoColorCode, Nunc™
Supplier: Thermo Fisher Scientific
Measuring scoops
Supplier: KARTELL
96- and 384-well cell culture microplates, Advanced TC™
Supplier: Greiner Bio-One
VWR®, Strips of PCR Tubes and Caps
Supplier: VWR Collection
Stoppers for tubes
Supplier: Greiner Bio-One
Storage Boxes
Supplier: EPPENDORF
Industrial scoops, Plastilab®
Supplier: KARTELL
Collagen I-coated microplates, Corning® BioCoat®
Supplier: Corning
Petri dishes
Supplier: DELTALAB