PIN REMOVAL KEY (REPLACED PR17) CLNPX2 1 * 5 L
Catalog # MLDVX4948
Supplier: Molecular Devices
Frequently Bought Together
VWR® Counter-Pen™ Cell Counter
Supplier: VWR Collection
Accessories for OxiTop® and OxiTop® Control
Supplier: WTW
ThermoMixer® C Thermomixer
Supplier: EPPENDORF
Single gas detectors, BW Clip
Supplier: BW TECHNOLOGIES
Guillotine tubing cutter
Supplier: VICI JOUR RESEARCH
Accessories for VWR® V-1200, UV-1600PC, UV-3100PC
Supplier: COLLECTION PREFIX HPLC
Flangeless fittings for ¹/₁₆" Ø tubing, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
Accessory for airflow indicator
Supplier: Dräger
Accessories for Mettler balances
Supplier: Mettler - Toledo
RDO SENSOR CAP REPLACEMENT 1 * 1 items
Supplier: Thermo Orion
VWR® pH CO 2500L, pH/Conductivity/°C Bench Meter with Colour Graphic Touch Screen
Supplier: VWR Collection
pH/mV/°C meters, bench, inoLab® pH 7110 / 7310
Supplier: WTW
Accessories for serial printers for eletrochemical meters
Supplier: CONSORT
Accessories for HygroPalm
Supplier: ROTRONIC INSTRUMENTS
Conductivity standards
Supplier: WTW
VWR® PH 20 pH Meter, Handheld
Supplier: VWR Collection
VWR® pHenomenal® MU 6100 H Multi-Parameter Meter (pH/Conductivity/DO), Handheld
Supplier: VWR Collection
Melting point and boiling point capillaries and tubes
Supplier: BUCHI
Combined titration electrodes, plug and play
Supplier: Mettler - Toledo