ContactUsLinkComponent
1 component: BDE 47 [CAS:5436-43-1] 50mg/l in Iso-octane CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store in a refrigerator at temperatures between 2°C to 8°C 1 * 1 mL
Catalog # CMPLRD0144291
1 component: BDE 47 [CAS:5436-43-1] 50mg/l in Iso-octane CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store in a refrigerator at temperatures between 2°C to 8°C 1 * 1 mL
Catalog # CMPLRD0144291
Frequently Bought Together