1 component: PBB 153 [CAS:59080-40-9] 35mg/l in Iso-octane CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store in a refrigerator at temperatures between 2°C to 8°C 1 * 1 mL
Catalog # CMPLRD0916131
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together