Order Entry
Belgium
Orders LinkContactUsLinkComponent
Ammonia solution

Ammonia solution

200 L
Catalog # SPCMAM180-200LTBL
Supplier:   Spectrum Chemical
Ammonia solution
Catalog # SPCMAM180-200LTBL
Supplier:  Spectrum Chemical
CAS Number:  1336-21-6
Ammonia solution

200 L

CAS Number:  1336-21-6
Price
3 990,00 €
Log in for availability
1 * 200 L
Product Details & Documents

Strong Ammonia Solution, NF has many uses such as in topical pharmaceutical solutions destined for treating insect bites. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol

Documents

Specifications

Specifications

  • Pk:
    200 L

Specifications