3 components: 100mg/l each of Chlorates (ClO3-) Oxalates (C2O42-) Chlorites (ClO2-) in Water 1 * 100 mL
N° de catalogue CMPLRM090972L1
Supplier: CUSTOM MADE CHEMICALS LAB
3 components: 100mg/l each of Chlorates (ClO3-) Oxalates (C2O42-) Chlorites (ClO2-) in Water 1 * 100 mL
N° de catalogue CMPLRM090972L1
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Sodium hydroxide ≥98%, pellets, GPR RECTAPUR®
Supplier: VWR Chemicals
VWR®, Test strips, chloride, Dosatest
Supplier: VWR Chemicals
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Ammonia solution ∼32%, GPR RECTAPUR®
Supplier: VWR Chemicals
Test kits, ammonium, Spectroquant®, Supelco®
Supplier: Merck
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Test kits, surfactants, Spectroquant®, Supelco®
Supplier: Merck
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Test strips, peroxide, QUANTOFIX®
Supplier: MACHEREY-NAGEL
Sodium hydroxide 97.0-100.5%, pellets NF, JP, Ph. Eur., BP
Supplier: VWR Chemicals
Acid red 27 ≥85-95% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Methylene blue
Supplier: RAL
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach
4 COMPONENTS MIX 1 * 100 mL
Supplier: CUSTOM MADE CHEMICALS LAB