ContactUsLinkComponent
6 components: 100mg/l each of Ge In Lu Rh Sc Tb in HNO3 1% HCl 5% ,, CRM, ISO 17034 and ISO 17025,, Conservation 24 months. (12 months after opening),, Ambient Temp.,, 1 * 100 mL
Catalog # CMPLRM065086L1
6 components: 100mg/l each of Ge In Lu Rh Sc Tb in HNO3 1% HCl 5% ,, CRM, ISO 17034 and ISO 17025,, Conservation 24 months. (12 months after opening),, Ambient Temp.,, 1 * 100 mL
Catalog # CMPLRM065086L1
Frequently Bought Together