SODIUM HYDROXIDE SOLUTION 0.1N BIOCERT 1 * 5 L
SPCMS9291-5LT
Frequently Bought Together
Formalin 10%, Q Path®, buffered
: VWR Chemicals
Acid red 27 ≥85-95% (dye content), Sigma-Aldrich®
: SIGMA ALDRICH MICROSCOPY
Ammonium acetate, Supelco®
: Merck
trans-4-Hydroxycinnamic acid ≥98% (by HPLC), Sigma-Aldrich®
: SIGMA ALDRICH MICROSCOPY
Urine test strips, Medi-Test
: MACHEREY-NAGEL