Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Utility carts

Cleanroom utility cart with casters, 865×550×990 mm, 2
  PALBUC-85-2-ESD
 :   Palbam
Utility carts
  PALBUC-85-2-ESD
 :  Palbam
Utility carts
Utility carts

Cleanroom utility cart with casters, 865×550×990 mm, 2

 
2 240,00 €
   
1 * 1 ST
 

These cleanroom carts are designed to withstand heavy loads.

  • Up to 60 kg per shelf
  • Integral raised ledges prevent parts from rolling off and are easy to wipe down
  • Choice of two or three shelves
  • Solid or perforated
  • White nylon 130 mm (5") cleanroom castors (Two with brakes) supplied with every cart

Stainless steel 304 with electropolished finish. Special non-marking, smooth running.

 : Typically available from stock for direct shipment from the supplier. Shipped flat-packed with assembly instructions.

 
  • Description:
    Cleanroom utility cart with casters
  • W×D×H:
    865×550×990 mm
  • Shelves:
    2