2-(1-METHYL-5-OXOPYRROLIDIN-3-YL)ACETICACID 1 * 25 g
Catalog # PHBLPBZ6042-25G
Supplier: Pharmablock (USA)
2-(1-METHYL-5-OXOPYRROLIDIN-3-YL)ACETICACID 1 * 25 g
Catalog # PHBLPBZ6042-25G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethyl acetate ≥99.9%, PESTINORM® Supra Trace for organic trace analysis
Supplier: VWR Chemicals
1-Propanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Pyrene for synthesis, Sigma-Aldrich®
Supplier: Merck
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Butyl acetate ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
tert-Butyl methyl ether 99%, pure
Supplier: Thermo Fisher Scientific
Xylene (mixture of isomers), extra dry 98+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Tetrahydrofuran, extra dry 99.85% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
n-Pentane ≥99% for HPLC
Supplier: Thermo Fisher Scientific
Glycerine ≥99%
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal®
Supplier: Thermo Fisher Scientific
2-Propanol 99.5% for HPLC
Supplier: Thermo Fisher Scientific
Methanol, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
2-Propanol ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
1-Hexanol 98%, Reagent Grade
Supplier: Honeywell Chemicals
2-Propanol, BAKER, Laboratory Reagent, J.T.Baker®
Supplier: Avantor