ContactUsLinkComponent
Tpx hose connector, straight, transparent. in very strong performance with long, conical connections for large tolerance ranges. temperature resistant up to +130 ° c, sterilized 1 * 1 items
Catalog # SEMA4666
Supplier:  SEMADENI
Tpx hose connector, straight, transparent. in very strong performance with long, conical connections for large tolerance ranges. temperature resistant up to +130 ° c, sterilized 1 * 1 items
Catalog # SEMA4666
Supplier:  SEMADENI
Frequently Bought Together