Order Entry
Belgium
ContactUsLinkComponent
Masterflex® Ismatec® IPC Peristaltic Pump, 0.1 to 11.25 rpm, 12-Channel, 115/230 VAC, Avantor®
Masterflex® Ismatec® IPC Peristaltic Pump, 0.1 to 11.25 rpm, 12-Channel, 115/230 VAC, Avantor®
Catalog # MFLX78006-44-EU
undefined
Masterflex® Ismatec® IPC Peristaltic Pump, 0.1 to 11.25 rpm, 12-Channel, 115/230 VAC, Avantor®
Catalog # MFLX78006-44-EU

About this item

Achieve high-accuracy multi-channel flow, with an intuitive touch-screen interface.

  • Replicate same flow for all channels; perfect for auto-analysis, reactor studies, cell line seeding, and research
  • Planetary drive with 8 stainless steel rollers reduces pulsation, increases flow rate accuracy, and lengthens tubing life
  • 12,7 cm (5") capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
  • Flexible operational control via screen or remote control (analog remote control, serial, or EtherNet/IP)
  • Three-level user access – gives you the option of controlling who can operate the pump and at what level
  • Pump lets you save and retrieve commonly used dispense programs for rapid set up
  • Continuous run, volume dispense, and time dispense modes; batch control and interval/off time in volume and time modes

These Ismatec® IPC multichannel pumps feature planetary gears that drive eight stainless steel rollers in unison for smooth rotation and accurate liquid flow. The intuitive touchscreen display shows flow rate or RPM (min⁻¹), allows for easy calibration, features configurable volume or time dispense modes, and lets you save and recall up to 50 programs. Volumetric dispensing features a unique roller positioning system to ensure repeatability.

Mix and match tubing sizes and formulations across the channels to suit your application. Included Click’n’go™ cartridges feature automatic occlusion for simplified use, fewer tubing changes, and reproducible results. Optional pressure lever cartridges offer you the opportunity to manually adjust the back pressure experienced by the tubing / pump combination (order separately).

Pumps feature a variety of built in options for remote communications. EtherNet/IP control is available via RJ45. RS-232 serial control is available via a DB9 interface or USB port (USB use requires cable MFLX78006-90). A 25-pin connector allows analog input and output controls.

Certifications: UL, cUL, CE, ISO9001, RoHS, REACH.
Ordering information: Order the two-stop tubing size and formulation to suit your application (sold separately). More than 100 options exist — the most popular sizes and formulations can be found on the accessory tab.
Delivery: Includes pump, power adapter with 1,9 m (6') cable, and click n'go cartridges (order tubing separately).
  • Power:
    90 - 260 VAC; 50/60 Hz
  • Accuracy:
    ±1,0%
  • Cloud connected:
    Yes
  • Control type:
    Digital
  • Frequency:
    50/60 Hz
  • Max. pressure:
    1 bar (14,5 psi)
  • Min. flow rate:
    0,0004 ml /min
  • No. of channels:
    12
  • Voltage:
    90 - 260 VAC
  • Description:
    Masterflex® Ismatec® IPC peristaltic pump, 0.1 - 11.25 rpm, 12-channel, 115/230 VAC
  • Height:
    200 mm
  • Length:
    390 mm
  • Seq:
    16
  • Environmentally Preferable:
  • Model:
    78006-44
  • Duty cycle:
    Continuous
  • Anti-drip:
    Yes
  • Digital dispensing:
    Yes
  • Drive IP rating:
    IP 31
  • Max. flow rate per channel:
    11 ml/min
  • Min. flow rate per channel:
    0,0004 ml /min
  • Min. RPM:
    0,11 rpm (min⁻¹)
  • Open-head sensor:
    No
  • Remote I/O:
    EtherNet/IP, Serial RS-232, MasterflexLive, 0 - 20 mA, 4 - 20 mA, 0 - 10 V, start/stop, direction, and prime via 25 pin contact closure
  • Reversible motor:
    Yes
  • Tubing sizes accepted:
    Two-stop 0,13 - 3,17 mm
  • Washdown capable:
    No
  • Drive:
    IPC 11
  • No. of rollers:
    8
  • Warranty:
    2 years
  • Series:
    Ismatec
  • Pk:
    1
  • Width:
    193 mm
  • Electrical plug type:
    EU
Frequently Bought Together