Order Entry
Belgium
Orders LinkContactUsLinkComponent
Conductive Tote Boxes

Conductive Tote Boxes

Conductive tote box, 600×400×236 mm
  DESC239345
 :   DESCO
Conductive Tote Boxes
  DESC239345
 :  DESCO
Conductive Tote Boxes

Conductive tote box, 600×400×236 mm

 
69,90 €
   
1 * 1 ST
 

Conductive tote boxes are made from injection moulded conductive polypropylene.

  • Flat base, stackable
  • Rv: 1×10³ to <1×10⁶ Ω
  • Custom-made dividers are available for PCB containers
  • Machined from approximately 2 mm thick conductive plastic or Cortronic® B-flute static dissipative cardboard
  • Many shapes and sizes possible
  • Process is suited for small and medium quantities
 
  • Description:
    Conductive tote box, 600×400×236 mm
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.