Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Conductive Tote Boxes

Conductive tote box, 400×300×120 mm
  DESC239305
 :   DESCO
Conductive Tote Boxes
  DESC239305
 :  DESCO
Conductive Tote Boxes

Conductive tote box, 400×300×120 mm

 
27,40 €
   
1 * 1 ST
 

Conductive tote boxes are made from injection moulded conductive polypropylene.

  • Flat base, stackable
  • Rv: 1×10³ to <1×10⁶ Ω
  • Custom-made dividers are available for PCB containers
  • Machined from approximately 2 mm thick conductive plastic or Cortronic® B-flute static dissipative cardboard
  • Many shapes and sizes possible
  • Process is suited for small and medium quantities
 
  • Description:
    Conductive tote box, 400×300×120 mm
  • Seq:
    01
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.