Orders LinkContactUsLinkComponent
O-Rings, Graphite
O-Rings, Graphite
Graphite O-ring for Shimadzu 17A, 2010 and 2014 splitless
Catalog # SGEA216639
O-Rings, Graphite
Catalog # SGEA216639
O-Rings, Graphite
Graphite O-ring for Shimadzu 17A, 2010 and 2014 splitless
Price
49,10 €
Log in for availability
1 * 10 ST
Product Details & Documents
Specifications
  • Description:
    Graphite O-ring for Shimadzu 17A, 2010 and 2014 splitless
  • Seq:
    03