((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate,95% 1 * 5 g
Catalog # AMBDA160197-5G
Supplier: AMBEED
((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate,95% 1 * 5 g
Catalog # AMBDA160197-5G
Supplier: AMBEED
Frequently Bought Together
Benzyl acetate for synthesis, Sigma-Aldrich®
Supplier: Merck
Isoamyl alcohol 98%, pure
Supplier: Thermo Fisher Scientific
Methanol ≥99.9% ACS, Spectrophotometric Grade for spectrophotometry, Riedel-de Haën™
Supplier: Honeywell Chemicals
N,N-Dimethylformamide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide, extra dry over Molecular Sieve 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Supplier: Merck
Acetonitrile Reag. Ph. Eur., gradient grade for HPLC, Burdick & Jackson™
Supplier: Honeywell Chemicals
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
Acetone, BAKER, J.T.Baker®
Supplier: Avantor
Isoamyl alcohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Bromoform 99+% stabilised
Supplier: Thermo Fisher Scientific