MIX HPLC EAU/ACÉTONITRILE 80%/20% 1 * 2,5 L
Catalog # PRLS90259.320
Supplier: PROLABO CMP
Frequently Bought Together
Embedding and mounting media
Supplier: Merck
Test kits, iron, Spectroquant®, Supelco®
Supplier: Merck
Schiff reagent for analysis of aldehydes
Supplier: VWR Chemicals
TAMPON PHOSPHATE PH 5.8 EUR. PH. 1 * 1 L
Supplier: CUSTOM MADE CHEMICALS LAB
DENARASE® highly active endonuclease from Serratia marcescens
Supplier: C LECTA
Acid Red 183 30% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
BIOTECTOR 1.8N SCHWEFELSAURE 80MG/L MN
Supplier: Hach
Powder pillows, manganese
Supplier: Hach
Reagents for Sulphide Determination using Lovibond® MD photometers
Supplier: Lovibond
CORROSION NF IN ISO 3651-2 + A 262 PR E contains H2SO4 and CuSO4,5 H2O 1 * 2,5 L
Supplier: PROLABO CMP