ContactUsLinkComponent
Specifications:- Capillary Column Phase: HP-5ms Ultra Inert. Capillary Tubing: Fused Silica. Film Thickness: 1 um. GC Column Format: LTM. GC Column Type: Ultra Inert. Inner Diameter (ID): 0.32 mm. Length: 30 m. Polarity: Low Polarity. With Smart Key 1 * 1 Pce
Catalog # HEWL19091S-213UILT
Supplier:  Agilent
Specifications:- Capillary Column Phase: HP-5ms Ultra Inert. Capillary Tubing: Fused Silica. Film Thickness: 1 um. GC Column Format: LTM. GC Column Type: Ultra Inert. Inner Diameter (ID): 0.32 mm. Length: 30 m. Polarity: Low Polarity. With Smart Key 1 * 1 Pce
Catalog # HEWL19091S-213UILT
Supplier:  Agilent
Frequently Bought Together