ContactUsLinkComponent
Specifications:- Capillary Column Phase: Select Low Sulfur. Capillary Tubing: Fused Silica. GC Column Dim: 60 m x . GC Column Format : Intuvo. GC Column Type: PLOT. Inner Diameter (ID): 0.32 mm. Length: 60 m. Length (m): 60 m. With Smart Key: Yes. 1 * 1 items
Catalog # HEWLCP8575-INT
Supplier:  Agilent
Specifications:- Capillary Column Phase: Select Low Sulfur. Capillary Tubing: Fused Silica. GC Column Dim: 60 m x . GC Column Format : Intuvo. GC Column Type: PLOT. Inner Diameter (ID): 0.32 mm. Length: 60 m. Length (m): 60 m. With Smart Key: Yes. 1 * 1 items
Catalog # HEWLCP8575-INT
Supplier:  Agilent
Frequently Bought Together