ContactUsLinkComponent
Specifications:- Capillary Column Phase: CP-Sil 5 CB. Capillary Tubing: Fused Silica. Film Thickness: 5 um. GC Column Dim: 50 m x 5 um. GC Column Format: Intuvo. Inner Diameter (ID): 0.32 mm. Length: 50 m. Length (m): 50 m. Polarity: Low Polarity. Wi 1 * 1 items
Catalog # HEWLCP7690-INT
Supplier:  Agilent
Specifications:- Capillary Column Phase: CP-Sil 5 CB. Capillary Tubing: Fused Silica. Film Thickness: 5 um. GC Column Dim: 50 m x 5 um. GC Column Format: Intuvo. Inner Diameter (ID): 0.32 mm. Length: 50 m. Length (m): 50 m. Polarity: Low Polarity. Wi 1 * 1 items
Catalog # HEWLCP7690-INT
Supplier:  Agilent
Frequently Bought Together