ContactUsLinkComponent
Specifications:- Capillary Column Phase: Select PAH. Capillary Tubing: Fused Silica. Film Thickness: 0.15 um. GC Column Dim: 30 m x 0.15 um. GC Column Format: Intuvo. Inner Diameter (ID): 0.25 mm. Length: 30 mm. Length (m): 30 m. Polarity: Low Polari 1 * 1 items
Catalog # HEWLCP7462-INT
Supplier:  Agilent
Specifications:- Capillary Column Phase: Select PAH. Capillary Tubing: Fused Silica. Film Thickness: 0.15 um. GC Column Dim: 30 m x 0.15 um. GC Column Format: Intuvo. Inner Diameter (ID): 0.25 mm. Length: 30 mm. Length (m): 30 m. Polarity: Low Polari 1 * 1 items
Catalog # HEWLCP7462-INT
Supplier:  Agilent
Frequently Bought Together