ContactUsLinkComponent
Specifications:- Capillary Column Phase: HP-5ms. Capillary Tubing: Fused Silica. Film Thickness: 0.18 um. GC Column Format: Intuvo. GC Column Type: GC/MS. Inner Diameter (ID): 0.18 mm. Length: 20 m. Polarity: Low Polarity. With Smart Key: Yes. 1 * 1 Pce
Catalog # HEWL19091S-577UI-I
Supplier:  Agilent
Specifications:- Capillary Column Phase: HP-5ms. Capillary Tubing: Fused Silica. Film Thickness: 0.18 um. GC Column Format: Intuvo. GC Column Type: GC/MS. Inner Diameter (ID): 0.18 mm. Length: 20 m. Polarity: Low Polarity. With Smart Key: Yes. 1 * 1 Pce
Catalog # HEWL19091S-577UI-I
Supplier:  Agilent
Frequently Bought Together