ContactUsLinkComponent
Specifications:- Capillary Column Phase: HP-5ms. Capillary Tubing: Fused Silica. Film Thickness: 1 um. GC Column Format: Intuvo. GC Column Type: GC/MS. Inner Diameter (ID): 0.32 mm. Length: 30 m. Polarity: Low Polarity. With Smart Key: Yes. 1 * 1 Pce
  HEWL19091S-213UI-I
 :  Agilent
Specifications:- Capillary Column Phase: HP-5ms. Capillary Tubing: Fused Silica. Film Thickness: 1 um. GC Column Format: Intuvo. GC Column Type: GC/MS. Inner Diameter (ID): 0.32 mm. Length: 30 m. Polarity: Low Polarity. With Smart Key: Yes. 1 * 1 Pce
  HEWL19091S-213UI-I
 :  Agilent
Frequently Bought Together