ContactUsLinkComponent
Specifications:- Capillary Column Phase: DB-5ms Ultra Inert. Capillary Tubing: Fused Silica. Film Thickness: 0.5 um. GC Column Forma t: Intuvo. GC Column Type: Ultra Inert. Inner Diameter (ID): 0.25 mm. Length: 30 m. Polarity: Low Polarity. With Smar 1 * 1 items
Catalog # HEWL122-5536UI-INT
Supplier:  Agilent
Specifications:- Capillary Column Phase: DB-5ms Ultra Inert. Capillary Tubing: Fused Silica. Film Thickness: 0.5 um. GC Column Forma t: Intuvo. GC Column Type: Ultra Inert. Inner Diameter (ID): 0.25 mm. Length: 30 m. Polarity: Low Polarity. With Smar 1 * 1 items
Catalog # HEWL122-5536UI-INT
Supplier:  Agilent
Frequently Bought Together