ContactUsLinkComponent
Specifications:- Capillary Column Phase: DB-17ms. Capillary Tubing: Fused Silica. Film Thickness: 0.25 um. GC Column Format: Intuvo. GC Column Type: Low Bleed. Inner Diameter (ID): 0.25 mm. Length: 30 m. Polarity: Mid Polarity. With Smart Key: Yes. 1 * 1 items
Catalog # HEWL122-4732-INT
Supplier:  Agilent
Specifications:- Capillary Column Phase: DB-17ms. Capillary Tubing: Fused Silica. Film Thickness: 0.25 um. GC Column Format: Intuvo. GC Column Type: Low Bleed. Inner Diameter (ID): 0.25 mm. Length: 30 m. Polarity: Mid Polarity. With Smart Key: Yes. 1 * 1 items
Catalog # HEWL122-4732-INT
Supplier:  Agilent
Frequently Bought Together